Iron(II) sulphate hydrate, getrocknet, Pulver USP

Lieferant: Spectrum Chemical
Achtung

F1065-2.5KG
SPCMF1065-2.5KGEA 582 EUR
SPCMF1065-2.5KG
Iron(II) sulphate hydrate, getrocknet, Pulver USP
Eisen(II)sulfat Hydrat

Dried Ferrous Sulfate, Powder, USP is a mineral supplement. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: FeSO₄·H₂O
Schmelzpunkt: ∼300 °C
Dichte: 2,97 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149722
CAS-Nummer: 13463-43-9
EINECS: 231-753-5

Bestellen

Spezifikation Testergebnisse

Assay 86% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR