Indigocarmin

Lieferant: Spectrum Chemical
Achtung

Synonyme: Acid Blue 74, Food blue No. 2

FD115-100GM FD115-25GM FD115-500GM
SPCMFD115-100GMEA 184 EUR
SPCMFD115-100GM SPCMFD115-25GM SPCMFD115-500GM
Indigocarmin
Indigocarmin

FD and C Blue No. 2 - In the United States, the Food and Drug Administration (FDA) controls the specifications for Food, Drug and Cosmetic (FD&C) colours.


Spectrum certifies all batches of FD&C colours meet the FDA's quality standards.


Bitte beachten Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.

Formel: C₁₆H₈N₂Na₂O₈S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00005723
CAS-Nummer: 860-22-0

Bestellen

Spezifikation Testergebnisse

Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR