Glycerine (synthetisch) ≥99% BP, Ph. Eur., JP, USP
Lieferant: Spectrum Chemical
Synonyme:
1,2,3-Propantriol, Glycerol
G1015-4LTPL
G1015-20LTPL
G1015-200LTBL
SPCMG1015-4LTPLEA
0
EUR
SPCMG1015-4LTPL
SPCMG1015-20LTPL
SPCMG1015-200LTBL
Glycerine (synthetisch) ≥99% BP, Ph. Eur., JP, USP
Glycerin
Glycerin, Synthetic, USP, EP, BP, JP is one of two USP grade Glycerins available from Spectrum with the other being Glycerin, Natural, USP, EP, BP. For those products requiring FDA approvals, if the initial formulation was done with Glycerin, Synthetic, USP, EP, BP, JP, the use of Glycerin, Natural, USP, EP, BP, JP is not interchangeable but would require the new formulation to undergo a new FDA approval process.
Formel:
HOCH₂CH(OH)CH₂OH Siedepunkt: 290 °C (1013 hPa) Schmelzpunkt: 18,6 °C Dichte: 1,264 Flammpunkt: 177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004722 CAS-Nummer: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Spezifikation Testergebnisse
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...