L(+)-Glutamin 98,5 FCC
Lieferant: Spectrum Chemical
Synonyme:
L(+)-Glutaminsäure-5-amid, Q, (S)-(+)-2-Amino-1,5-pentandisäure-5-amid, (S)-(+)-Glutamin, L(+)-2-Amino-1,5-pentandisäure-5-amid, H-Gln-OH, (S)-(+)-Glutaminsäure-5-amid, Gln, Levoglutamid
L-Glutamine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₅H₁₀N₂O₃ Molecular Weight: 146.15 g/mol Siedepunkt: 355 °C (1013 hPa) Schmelzpunkt: 185…186 °C Dichte: 1.5326 g/cm³ (130 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008044 CAS-Nummer: 56-85-9 EINECS: 200-292-1 Merck Index: 13,04484 |
|
Spezifikation Testergebnisse
Assay | 98.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...