L(-)-Glutathione (reduced form) ≥96%, reduziert

Lieferant: Spectrum Chemical

Synonyme: L(-)-Glutathion, reduziert, γ-L-Glutamyl-L-cysteinylglycin, GSH

GL146-25GM
SPCMGL146-25GMEA 319 EUR
SPCMGL146-25GM
L(-)-Glutathione (reduced form) ≥96%, reduziert
L(-)-Glutathion (reduzierte Form)

L-Glutathione, Reduced can also be referred to as GSH and is an organic chemical that can be traced in plants and animals alike. It is an antioxidant that prevents damage from reactive oxygen species to cellular components that are important. It can be used to raise levels of plasma and liver GSH concentrations and is thought to have some positive effects on reducing cancer development.

Formel: C₁₀H₁₇N₃O₆S
Molecular Weight: 307,33 g/mol
Schmelzpunkt: 185…195 °C
Dichte: 1,475 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00065939
CAS-Nummer: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488

Bestellen

Spezifikation Testergebnisse

Assay 96% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR