Hydroxyurea ≥97% USP

Lieferant: Spectrum Chemical

Synonyme: Hydroxycarbamid, Aminoameisensäurehydroxyamid

H1181-12KGBL H1181-25KGBL H1181-500GM H1181-25GM H1181-50GM
SPCMH1181-12KGBLEA 0 EUR
SPCMH1181-12KGBL SPCMH1181-25KGBL SPCMH1181-500GM SPCMH1181-25GM SPCMH1181-50GM
Hydroxyurea ≥97% USP
Hydroxyharnstoff

Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: CH₄N₂O₂
Molecular Weight: 76.05 g/mol
Siedepunkt: 296 °C (1013 hPa)
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007943
CAS-Nummer: 127-07-1
EINECS: 204-821-7
Merck Index: 13,04874

Bestellen

Spezifikation Testergebnisse

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR