Hydroxyurea ≥97% USP
Lieferant: Spectrum Chemical
Synonyme:
Hydroxycarbamid, Aminoameisensäurehydroxyamid
H1181-12KGBL
H1181-25KGBL
H1181-500GM
H1181-25GM
H1181-50GM
SPCMH1181-12KGBLEA
0
EUR
SPCMH1181-12KGBL
SPCMH1181-25KGBL
SPCMH1181-500GM
SPCMH1181-25GM
SPCMH1181-50GM
Hydroxyurea ≥97% USP
Hydroxyharnstoff
Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
CH₄N₂O₂ Molecular Weight: 76.05 g/mol Siedepunkt: 296 °C (1013 hPa) Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007943 CAS-Nummer: 127-07-1 EINECS: 204-821-7 Merck Index: 13,04874 |
Spezifikation Testergebnisse
Assay | 97% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...