Bole white (kaolin), Pulver FCC

Lieferant: Spectrum Chemical

Synonyme: Bolus Alba, Porzellanerde, Porzellanton, hydratisiertes Aluminiumsilikat, Pfeifenerde

K1004-500GM K1004-2.5KG
SPCMK1004-500GMEA 145 EUR
SPCMK1004-500GM SPCMK1004-2.5KG
Bole white (kaolin), Pulver FCC
Weißer Ton (Kaolin)

Kaolin, Powder, FCC is used as a whitener in toothpaste. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: Al₂SiO₇·2H₂O
Molecular Weight: 258.16 g/mol
Schmelzpunkt: 1760
Lagertemperatur: Raumtemperatur
MDL: MFCD00062311
CAS-Nummer: 1332-58-7
Merck Index: 13,05300

Bestellen

Spezifikation Testergebnisse

Acid-Soluble Substances max. 2.0%
Arsenic (As) max.3 mg/kg
Carbonate (CO₃) To Pass Test
Iron (Fe) To Pass Test
Lead (Pb) max. 10 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR