Manganese(II) chloride tetrahydrate ≥98% USP

Lieferant: Spectrum Chemical
Achtung

Synonyme: Mangandichlorid Tetrahydrat

M1106-500GM M1106-125GM M1106-2.5KG
SPCMM1106-500GMEA 428 EUR
SPCMM1106-500GM SPCMM1106-125GM SPCMM1106-2.5KG
Manganese(II) chloride tetrahydrate ≥98% USP
Mangan(II)chlorid Tetrahydrat

Manganese Chloride, Tetrahydrate, USP is indicated for use as a supplement to intravenous solutions given for total parenteral nutrition (TPN) to maintain Manganese serum levels. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: MnCl₂·4H₂O
Siedepunkt: 1190 °C (1013 hPa)
Schmelzpunkt: 58…59 °C
Dichte: 2,01 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149792
CAS-Nummer: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 13,05751

Bestellen

Spezifikation Testergebnisse

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR