Mineral oil light FCC, niedrige Viskosität
Lieferant: Spectrum Chemical
Mineral Oil, Low Viscosity, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00131611 CAS-Nummer: 8042-47-5 |
Spezifikation Testergebnisse
Carbon Number at 5% Distillation Point | 17 - 25 |
Specific Gravity @ 25°C | 0.829 - 0.859 |
Ultraviolet Absorbance (Polynuclear Hydrocabons) | To Pass Test |
Viscosity @ 100°C | 3 - 11 cSt |
Lead (Pb) | max. 1 mg/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...