Dimethyl sulphone ≥99%, Kristalle
Lieferant: Spectrum Chemical
Synonyme:
Methyl sulphone, Dimethylsulphone
M2125-500GM
SPCMM2125-500GMEA
395
EUR
SPCMM2125-500GM
Dimethyl sulphone ≥99%, Kristalle
Dimethylsulfon
Methyl Sulfone, Crystal is also known as sulfonylbismethane or methylsulfonylmethane. It is primarily used as a solvent for organic and inorganic substances because of its polarity and thermal stability.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₂H₆O₂S Molecular Weight: 94,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007566 CAS-Nummer: 67-71-0 |
Spezifikation Testergebnisse
Assay | 0.99 |
Melting Point | 107°C min. |
Loss On Drying | max. 1 % |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...