Weißes Mineralöl (Erdöl) leicht NF

Lieferant: Spectrum Chemical
Achtung

MI501-1LT MI501-20LTPL MI501-4LTGL
SPCMMI501-1LTEA 196 EUR
SPCMMI501-1LT SPCMMI501-20LTPL SPCMMI501-4LTGL
Weißes Mineralöl (Erdöl) leicht NF
Mineral oil, white (petroleum)

Light Mineral Oil, NF is used as a vehicle and humectant. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Siedepunkt: 300 °C (1013 hPa)
Dichte: 0,872 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00131611
CAS-Nummer: 8042-47-5
EINECS: 232-455-8

Bestellen

Spezifikation Testergebnisse

SPECIFIC GRAVITY @ 25°C 0.818-0.880
VISCOSITY @40.0 °C 3.0-34.4 mm²/s
ACIDITY TO PASS TEST
READILY CARBONIZABLE SUBSTANCES TO PASS TEST
POLYCYCLIC AROMATIC HYDROCARBONS TO PASS TEST
SOLID PARAFFIN TO PASS TEST
SULFUR COMPOUNDS TO PASS TEST
ELEMENTAL IMPURITIES AS REPORTED
IDENTIFICATION TO PASS TEST
RESIDUAL SOLVENTS TO PASS TEST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR