Phenol-Lösung 5% (w/v)

Lieferant: Spectrum Chemical
Gefahr

P-259-1LTPL P-259-4LTPL P-259-500ML
SPCMP-259-1LTPLEA 92.6 EUR
SPCMP-259-1LTPL SPCMP-259-4LTPL SPCMP-259-500ML
Phenol-Lösung 5% (w/v)
Phenol

Phenol, 5 Percent (w/v) Solution - Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₆H₅OH
MDL: MFCD00002143
CAS-Nummer: 108-95-2

Bestellen

Spezifikation Testergebnisse

Assay (w/v) 4.9-5.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR