Pluronic® L-61 (Synperonic® L 61) Tensid
Lieferant: Spectrum Chemical
Synonyme:
PEG-PPG-PEG, Synperonic® L61
Poloxamer 181, Surfactant is a nonionic triblock copolymer. It is made up of a main hydrophobic chain of polyoxypropylene bordered on each side by two hydrophilic chains of polyoxyethylene. These surfactants are commonly used in industrial applications, cosmetics, and pharmaceuticals. They have also been evaluated for various drug delivery applications and were shown to sensitise drug resistant cancers to chemotherapy. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formel:
HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a Molecular Weight: 2000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082049 CAS-Nummer: 9003-11-6 |
Spezifikation Testergebnisse
Cloud Point (10 % Aqueous) | 15 - 19 °C |
pH of a 2.5% Aqueous Solution | 5.0 - 7.5 |
Color (APHA) | max. 50 |
Water | max. 0.4 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...