Polysorbate 20 FCC

Lieferant: Spectrum Chemical

P1177-500MLGL
SPCMP1177-500MLGLEA 147 EUR
SPCMP1177-500MLGL
Polysorbate 20 FCC
Polysorbat 20

Polysorbate 20, FCC is used as a wetting agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Allergen statement (available upon request)

Formel: C₅₈H₁₁₄O₂₆
Dichte: 1,095
Lagertemperatur: Raumtemperatur
MDL: MFCD00165986
CAS-Nummer: 9005-64-5
EINECS: 500-018-3

Bestellen

Spezifikation Testergebnisse

Assay 70% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR