Kaliumcarbonat, wasserfrei ≥99,5%, körnig USP

Lieferant: Spectrum Chemical
Achtung

P1238-12KG P1238-2.5KG P1238-45KGBL
SPCMP1238-12KGEA 976 EUR
SPCMP1238-12KG SPCMP1238-2.5KG SPCMP1238-45KGBL
Kaliumcarbonat, wasserfrei ≥99,5%, körnig USP
Kaliumcarbonat

Potassium Carbonate, Anhydrous, Granular, USP is used as a drying agent for some non acidic compounds and as a buffering agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formel: K₂CO₃
Schmelzpunkt: 891 °C
Dichte: 2,42 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011382
CAS-Nummer: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR