Kaliumchlorid ≥99,0%, Kristalle FCC

Lieferant: Spectrum Chemical

Synonyme: Potassiumchloride

P1254-2.5KG P1254-500GM
SPCMP1254-2.5KGEA 390 EUR
SPCMP1254-2.5KG SPCMP1254-500GM
Kaliumchlorid ≥99,0%, Kristalle FCC
Potassium chloride

Potassium Chloride, Crystal, FCC is used as a flavor enhancer, flavoring agent, nutrient supplement, pH control agent, and stabiliser or thickener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formel: KCl
Molecular Weight: 74.55 g/mol
Siedepunkt: 1420
Schmelzpunkt: 773
Lagertemperatur: Raumtemperatur
MDL: MFCD00011360
CAS-Nummer: 7447-40-7
Merck Index: 14,07621

Bestellen

Spezifikation Testergebnisse

Assay (KCl; Dried Basis) 99.0 % min.
Acidity / Alkalinity To Pass Test
Heavy Metals (As Pb) max. 5 mg/kg
Iodide or Bromide To Pass Test
Loss On Drying max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR