Propionic acid ≥99,5% FCC

Lieferant: Spectrum Chemical
Gefahr

Synonyme: Methylessigsäure, Propansäure

P1326-4LTGL
SPCMP1326-4LTGLEA 738 EUR
SPCMP1326-4LTGL
Propionic acid ≥99,5% FCC
Propionsäure

Propionic Acid, FCC is used as a preservative and flavoring agent in packaged foods such as baked goods and cheese. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₃H₆O₂
Molecular Weight: 74,08 g/mol
Siedepunkt: 141 °C (1013 hPa)
Schmelzpunkt: –21 °C
Dichte: 0,993
Flammpunkt: 49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002756
CAS-Nummer: 79-09-4
EINECS: 201-176-3
UN: 3463
ADR: 8.3,II
REACH: 01-2119486971-24
Merck Index: 13,07917

Bestellen

Spezifikation Testergebnisse

Assay 99.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR