Kaliumdihydrogenphosphat ≥98,0% FCC

Lieferant: Spectrum Chemical

P1382-500GM
SPCMP1382-500GMEA 223 EUR
SPCMP1382-500GM
Kaliumdihydrogenphosphat ≥98,0% FCC
Kaliumdihydrogenphosphat

Potassium Phosphate Monobasic, FCC is used to stabilise, thicken and regulate the acidity and moisture in foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: KH₂PO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0

Bestellen

Spezifikation Testergebnisse

Assay (Dried Basis) 98.0 % min.
Arsenic (As) max.3 mg/kg
Fluoride max. 10 mg/kg
Insoluble Substances max. 0.2 %
Lead (Pb) max. 2 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR