Pluronic® L-61 (Synperonic® L 61)

Lieferant: Spectrum Chemical

Synonyme: Synperonic® L62, PEG-PPG-PEG, Pluronic® L-62

P1813-500ML
SPCMP1813-500MLEA 123 EUR
SPCMP1813-500ML
Pluronic® L-61 (Synperonic® L 61)
Pluronic® L-61 (Synperonic® L 61)

Poloxamer 182 belongs to a group of compounds known as poloxamers that are made up of three polymer blocks. In cosmetic and personal care product formulations, it functions as a surfactant - cleansing agent. It can be used in soaps and cleansers.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
Lagertemperatur: Raumtemperatur
MDL: MFCD00082049
CAS-Nummer: 9003-11-6

Bestellen

Spezifikation Testergebnisse

Cloud Point (10 % Aqueous) 22 - 26 °C
pH of a 2.5% Aqueous Solution 5.0- 7.5
Color (APHA) max. 50
Water max. 0.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR