D-Panthenol ≥98% FCC

Lieferant: Spectrum Chemical

Synonyme: Dexpanthenol, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

PA118-45KGBL
SPCMPA118-45KGBLEA 20330 EUR
SPCMPA118-45KGBL
D-Panthenol ≥98% FCC
D-Panthenol

Dexpanthenol, FCC is the provitamin of vitamin B5 and is used in nutritional supplements. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₉H₁₉NO₄
Molecular Weight: 205.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065006
CAS-Nummer: 81-13-0
EINECS: 201-327-3

Bestellen

Spezifikation Testergebnisse

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR