Phenol ≥99,0%, verschmolzenen Kristalle USP
Lieferant: Spectrum Chemical
|
|
Synonyme:
Phenylalkohol, Hydroxybenzol, Carbolsäure
PH120-1.5KGPL
PH120-25KGBL
PH120-500GMGL
SPCMPH120-1.5KGPLEA
636
EUR
SPCMPH120-1.5KGPL
SPCMPH120-25KGBL
SPCMPH120-500GMGL
Phenol ≥99,0%, verschmolzenen Kristalle USP
Phenol
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₆H₅OH Molecular Weight: 94.11 g/mol Siedepunkt: 181,7 Schmelzpunkt: 40,8 Dichte: 1.071 Flammpunkt: 81 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002143 CAS-Nummer: 108-95-2 UN: 1671 ADR: 6.1 II Merck Index: 12,07390 |
Spezifikation Testergebnisse
Assay (Anhydrous Basis) | 99.0 - 100.5 % |
Clarity of Solution and Reaction | To Pass Test |
Congealing Temperature | 39°C min. |
Water Determination | max. 0.5 % |
Nonvolatile Residue | max. 0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...