Polysorbate 20 NF
Lieferant: Spectrum Chemical
Synonyme:
Sorbitanmonolaurat, ethoxyliert, Sorbitanmonododecanoat, ethoxyliert, Polyoxyethylen-sorbitan-monolaurat, Polyethylenglycol-sorbitan-monolaurat
PO132-500MLGL
SPCMPO132-500MLGLEA
147
EUR
SPCMPO132-500MLGL
Polysorbate 20 NF
Polysorbat 20
Polysorbate 20, NF is a hydrophilic, nonionic, polysorbate surfactant and a polyoxyethylene derivative of sorbitan monolaurate with relative non-toxicity.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formel:
C₅₈H₁₁₄O₂₆ Dichte: 1,095 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00165986 CAS-Nummer: 9005-64-5 EINECS: 500-018-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...