Propylenglycol ≥99,5% USP
Lieferant: Spectrum Chemical
Synonyme:
Propylenglycol, (±)-1,2-Propylenglycol, (±)-1,2-Dihydroxypropan
PR130-20LTPL
PR130-4LTGL
PR130-4LTPL
PR130-200LTBL
PR130-500MLPL
SPCMPR130-20LTPLEA
1200
EUR
SPCMPR130-20LTPL
SPCMPR130-4LTGL
SPCMPR130-4LTPL
SPCMPR130-200LTBL
SPCMPR130-500MLPL
Propylenglycol ≥99,5% USP
Propylenglycol
Propylene Glycol, USP is used in pharmaceutical products as an excipient solvent in oral and topical drug products. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formel:
CH₃CH(OH)CH₂OH Molecular Weight: 76.1 g/mol Siedepunkt: 187.6 Schmelzpunkt: -59 Flammpunkt: 99 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064272 CAS-Nummer: 57-55-6 Merck Index: 13,07947 |
|
Spezifikation Testergebnisse
Assay | 99.5 % min. |
Specific Gravity @ 25°C | 1.035 - 1.037 |
Acidity | To Pass Test |
Water Determination | max. 0.2 % |
Residue On Ignition | max. 0.007 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...