Propylenglycol ≥99,5% USP

Lieferant: Spectrum Chemical

Synonyme: Propylenglycol, (±)-1,2-Propylenglycol, (±)-1,2-Dihydroxypropan

PR130-20LTPL PR130-4LTGL PR130-4LTPL PR130-200LTBL PR130-500MLPL
SPCMPR130-20LTPLEA 1200 EUR
SPCMPR130-20LTPL SPCMPR130-4LTGL SPCMPR130-4LTPL SPCMPR130-200LTBL SPCMPR130-500MLPL
Propylenglycol ≥99,5% USP
Propylenglycol

Propylene Glycol, USP is used in pharmaceutical products as an excipient solvent in oral and topical drug products. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formel: CH₃CH(OH)CH₂OH
Molecular Weight: 76.1 g/mol
Siedepunkt: 187.6
Schmelzpunkt: -59
Flammpunkt: 99
Lagertemperatur: Raumtemperatur
MDL: MFCD00064272
CAS-Nummer: 57-55-6
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 99.5 % min.
Specific Gravity @ 25°C 1.035 - 1.037
Acidity To Pass Test
Water Determination max. 0.2 %
Residue On Ignition max. 0.007 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR