Sodium hydroxide solution_SC 20% w/v

Lieferant: Spectrum Chemical

Synonyme: Caustic soda

S-229-500MLPL
SPCMS-229-500MLPLEA 61.9 EUR
SPCMS-229-500MLPL
Sodium hydroxide solution_SC 20% w/v
Sodium hydroxide

Sodium Hydroxide, 20 Percent (w/v) Solution, also known as caustic soda or lye, is used as a strong chemical base in the manufacture of textiles, drinking water, detergents, soaps and as a drain cleaner. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390
Schmelzpunkt: 323
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR