Sodium poly(naphthaleneformaldehyde)sulfonate
Lieferant: Spectrum Chemical
S1161-100GM
SPCMS1161-100GMEA
204
EUR
SPCMS1161-100GM
Sodium poly(naphthaleneformaldehyde)sulfonate
Sodium poly(naphthaleneformaldehyde)sulphonate
Synonyms: Blancol, naphthalenesulfonic acid, polymer with formaldehyde, sodium salt ; sodium salt of sulphonated naphthaleneformaldehyde condensate.
Formel:
C₁₀H₈O₃S·CH₂O)n·xNa |
CAS-Nummer:
9084-06-4 |
Spezifikation Testergebnisse
Appearance | Beige powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...