Trisodium citrate, wasserfrei ≥99,0%, Pulver FCC

Lieferant: Spectrum Chemical

S1254-2.5KG S1254-100GM S1254-500GM S1254-12KG
SPCMS1254-2.5KGEA 839 EUR
SPCMS1254-2.5KG SPCMS1254-100GM SPCMS1254-500GM SPCMS1254-12KG
Trisodium citrate, wasserfrei ≥99,0%, Pulver FCC
tri-Natriumcitrat

Sodium Citrate, Anhydrous, Powder, FCC is used as a flavorant and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₆H₅Na₃O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00012462
CAS-Nummer: 68-04-2

Bestellen

Spezifikation Testergebnisse

Assay (C₆H₅Na₃O₇; Anhydrous Basis) 99.0 - 100.5%
Lead max. 2 mg/kg
Alkalinity To Pass Test
Water Determination max. 1 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR