Sodium sesquicarbonate dihydrate
Lieferant: Spectrum Chemical
|
|
S1357-2.5KG
SPCMS1357-2.5KGEA
260
EUR
SPCMS1357-2.5KG
Sodium sesquicarbonate dihydrate
Sodium sesquicarbonate dihydrate
Sodium Sesquicarbonate, Dihydrate, with a systematic name trisodium hydrogen dicarbonate, is used as an alkalinity source for water treatment.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
Na₂CO₃·NaHCO₃·₂H₂O Molecular Weight: 226.03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036143 CAS-Nummer: 533-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...