Tetrasodium diphosphate decahydrate 99, crystals ACS

Lieferant: Spectrum Chemical
Achtung

Synonyme: TSPP, Natriumpyrophosphat tetrabasisch Decahydrat, tetra-Natriumpyrophosphat Decahydrat, Natriumdiphosphat Decahydrat, Natriumpyrophosphat Decahydrat

S1420-500GM
SPCMS1420-500GMEA 200 EUR
SPCMS1420-500GM
Tetrasodium diphosphate decahydrate 99, crystals ACS
tetra-Natriumdiphosphat Decahydrat

Sodium Pyrophosphate, Decahydrate, Crystal, Reagent, ACS is also known as tetrasodium pyrophosphate and is employed as a buffering agent as well as a thickening agent and can be found in many foods under the shape of a food additive. It is also used in toothpaste to aid in tartar control in the mouth. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formel: Na₄P₂O₇·10H₂O
Molecular Weight: 446.06 g/mol
Schmelzpunkt: 990 °C
Dichte: 1,82 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149200
CAS-Nummer: 13472-36-1
EINECS: 231-767-1

Bestellen

Spezifikation Testergebnisse

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR