Natriumhexadecylsulfat

Lieferant: Spectrum Chemical

Synonyme: Hexadecylschwefelsäure Natriumsalz, Hexadecylhydrogensulfat Natriumsalz, Schwefelsäurehexadecylester Natriumsalz

S1422-100GM
SPCMS1422-100GMEA 381 EUR
SPCMS1422-100GM
Natriumhexadecylsulfat
Natriumhexadecylsulfat

Sodium n-Hexadecyl sulfate is used as an anionic cleansing and emulsifying surfactant.


Ungraded products supplied by spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₁₆H₃₃NaO₄S
Molecular Weight: 344.49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047766
CAS-Nummer: 1120-01-0
EINECS: 214-292-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR