Stärke (aus Pfeilwurzel), Pulver

Lieferant: Spectrum Chemical

S1551-500GM
SPCMS1551-500GMEA 146 EUR
SPCMS1551-500GM
Stärke (aus Pfeilwurzel), Pulver
Starch

Starch, Arrowroot, Powder is used in talcum powders and hair dyes. It is added to moisturizers as a thickening agent and to help active ingredients penetrate the upper levels of the skin. This product is used in cosmetics to help moisturizers penetrate the skin. It appears as a fine, white powder. It may have other uses similar to cornstarch. In cosmetics, it is used as an abrasive.

Formel: (C₆H₁₀O₅)n
Molecular Weight: 162,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082026
CAS-Nummer: 9005-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR