Trisodium citrate, wasserfrei ≥99,0% USP

Lieferant: Spectrum Chemical

S1986-500GM S1986-2.5KG
SPCMS1986-500GMEA 252 EUR
SPCMS1986-500GM SPCMS1986-2.5KG
Trisodium citrate, wasserfrei ≥99,0% USP
tri-Natriumcitrat

Sodium Citrate, Anhydrous, USP is used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₆H₅Na₃O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00012462
CAS-Nummer: 68-04-2

Bestellen

Spezifikation Testergebnisse

Assay (Anhydrous Basis) 99.0 - 100.5%
Alkalinity To Pass Test
Water Determination max. 1.0 %
Tartrate To Pass Test
Heavy Metals max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR