Squalene ≥99%
Lieferant: Spectrum Chemical
Synonyme:
2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaen
S2002-5GLBL
SPCMS2002-5GLBLEA
7800
EUR
SPCMS2002-5GLBL
Squalene ≥99%
Squalen
Squalene, is an organic compound that be found occurring naturally in shark liver oil and certain plants. It is a hydrocarbon that all plants and animals produce in varying quantities. Mostly used in cosmetics as a natural skin moisturiser, it has recently been used as an immunologic adjuvant in certain vaccines.
Formel:
C₃₀H₅₀ Molecular Weight: 410,73 g/mol Siedepunkt: 429 °C (1013 hPa) Schmelzpunkt: –5 °C Dichte: 0,858 Flammpunkt: 200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008912 CAS-Nummer: 111-02-4 EINECS: 203-826-1 Merck Index: 13,08847 |
Spezifikation Testergebnisse
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...