di-Natriumhydrogenphosphat Heptahydrat ≥98,0%, Kristalle USP
Lieferant: Spectrum Chemical
Synonyme:
sek. Natriumphosphat Heptahydrat, Natriumphosphat dibasisch Heptahydrat
SO140-12KG
SPCMSO140-12KGEA
1250
EUR
SPCMSO140-12KG
di-Natriumhydrogenphosphat Heptahydrat ≥98,0%, Kristalle USP
di-Natriumhydrogenphosphat Heptahydrat
Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formel:
HNa₂O₄P·7H₂O Molecular Weight: 268,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149180 CAS-Nummer: 7782-85-6 |
Spezifikation Testergebnisse
Assay (Dried Basis) | 98.0 - 100.5 % |
Loss On Drying | 43.0 - 50.0 % |
Insoluble Matter | max. 0.4 % |
Chloride (Cl) | max. 0.06 % |
Sulfate (SO₄) | max. 0.2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...