Natriumchlorid

Lieferant: Spectrum Chemical

Synonyme: Saline, NaCl, Sea salt, Halite

SO155-500GM
SPCMSO155-500GMEA 106 EUR
SPCMSO155-500GM
Natriumchlorid
Natriumchlorid

Sodium Chloride, Granular, USP is used in a variety of pharmaceutical formulations, to produce isotonic formulations, in tablet formulations and as an osmotic agent. This product is not sterile and not intended for use in the manufacture of injectable dosage forms, peritoneal dialysis solutions, hemodialysis solutions or hemofiltration solutions.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: NaCl
Molecular Weight: 58.44 g/mol
Siedepunkt: 1413
Schmelzpunkt: 801
MDL: MFCD00003477
CAS-Nummer: 7647-14-5
Merck Index: 14,08599

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR