Sodium hydroxide 95,0, pellets NF

Lieferant: Spectrum Chemical

Synonyme: Caustic soda

SO170-500GM
SPCMSO170-500GMEA 115 EUR
SPCMSO170-500GM
Sodium hydroxide 95,0, pellets NF
Sodium hydroxide

Sodium Hydroxide, Pellets, NF is used as a pH adjusting agent.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390
Schmelzpunkt: 323
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

Assay (Total Alkali Including A 95.0 - 100.5%
Insoluble Substances & Organic Matter To Pass Test
Potassium (K) To Pass Test
Heavy Metals max. 30 ppm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR