Sodium iodide ≥99,0% USP

Lieferant: Spectrum Chemical

Synonyme: Iodnatrium

SO175-5GM
SPCMSO175-5GMEA 62.5 EUR
SPCMSO175-5GM
Sodium iodide ≥99,0% USP
Natriumiodid

Sodium Iodide, USP is a crystalline salt that is white and is used as a food supplement to counter and prevent iodine deficiencies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark;Health hazard

Formel: NaI
Molecular Weight: 149.89 g/mol
Siedepunkt: 1300
Schmelzpunkt: 662
Lagertemperatur: Raumtemperatur
MDL: MFCD00003532
CAS-Nummer: 7681-82-5
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Bestellen

Spezifikation Testergebnisse

Assay (NaI) 99.0 - 101.5%
Alkalinity To Pass Test
Water max. 2.0 %
Iodate (IO₃) To Pass Test
Nitrate, Nitrite, and Ammonia To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR