Natriumdihydrogenphosphat, wasserfrei ≥98,0% USP
Lieferant: Spectrum Chemical
Synonyme:
Natriumphosphat monobasisch
SO187-2.5KG
SPCMSO187-2.5KGEA
512
EUR
SPCMSO187-2.5KG
Natriumdihydrogenphosphat, wasserfrei ≥98,0% USP
Natriumdihydrogenphosphat
Monobasic Sodium Phosphate, Anhydrous, USP is used as a buffering agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formel:
NaH₂PO₄ Molecular Weight: 119,98 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003527 CAS-Nummer: 7558-80-7 |
Spezifikation Testergebnisse
Assay (NaH₂PO₄) | 98.0 - 103.0 % |
pH of a 5% Solution | 4.1 - 4.5 |
Water | max. 2.0 % |
Al, Ca, & Related Elements | To Pass Test |
Arsenic (As) | max. 8 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...