Soybean oil USP

Lieferant: Spectrum Chemical

SO255-200LTBL SO255-20LT SO255-4LTGL SO255-500MLGL
SPCMSO255-200LTBLEA 3180 EUR
SPCMSO255-200LTBL SPCMSO255-20LT SPCMSO255-4LTGL SPCMSO255-500MLGL
Soybean oil USP
Soybean oil

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Dichte: 0,917
Lagertemperatur: Raumtemperatur
MDL: MFCD00132356
CAS-Nummer: 8001-22-7

Bestellen

Spezifikation Testergebnisse

Alkaline impurities Passes test
Acid value Max. 0.3
Peroxide value Max. 10.0
Fatty acid composition Passes test
Heavy metals Max. 10 µg/g
Sterol composition (as Brassicasterol) Max. 0.3 %
Water Max. 0.1%
Unsaponifiable matter Max.1.5%
Identification Passes test
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR