Titanium(IV) oxide ≥99% USP
Lieferant: Spectrum Chemical
Titanium Dioxide, USP is used as a food coloring. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
TiO₂ Siedepunkt: 2900 °C (1013 hPa) Schmelzpunkt: 1855 °C Dichte: 4,2 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011269 CAS-Nummer: 13463-67-7 EINECS: 236-675-5 Merck Index: 13,09549 |
Spezifikation Testergebnisse
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...