Triacetin (glycerol triacetate) 97 USP
Lieferant: Spectrum Chemical
Synonyme:
Glycerintriacetat
TR106-500MLGL
SPCMTR106-500MLGLEA
206
EUR
SPCMTR106-500MLGL
Triacetin (glycerol triacetate) 97 USP
Triacetin (Glycerintriacetat)
Triacetin, USP is used as a humectant excipient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₉H₁₄O₆ Molecular Weight: 218.21 g/mol Siedepunkt: 258 °C (1013 hPa) Schmelzpunkt: 4 °C Dichte: 1,161 Flammpunkt: 138 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008716 CAS-Nummer: 102-76-1 EINECS: 203-051-9 Merck Index: 13,09664 |
Spezifikation Testergebnisse
Assay | 97% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...