n-Vinylpyrrolidone/Styrene copolymer 38%, wässrige Emulsion
Lieferant: Spectrum Chemical
N-Vinylpyrrolidone/Styrene Copolymer, 38 Percent Aqueous Emulsion, also known as N-Ethenyl-2-pyrrolidone, is an organic compound with a 5-membered lactam linked to a vinyl group. It is used as the precursor to polyvinylpyrrolidone.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Spezifikation Testergebnisse
pH | 2.0 - 5.0 |
Viscosity (25°C, BrooKField) | 200.0 - 800.0 cPs |
Solid Content | 38.0 - 42.0 % |