DL-α-Tocopherolacetat USP

Lieferant: Spectrum Chemical

Synonyme: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate

VI140-100ML
SPCMVI140-100MLEA 260 EUR
SPCMVI140-100ML
DL-α-Tocopherolacetat USP
DL-α-Tocopherolacetat

Vitamin E Acetate, USP is used as an antioxidant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₃₁H₅₂O₃
Molecular Weight: 472,75 g/mol
Dichte: 0,96
Lagertemperatur: Raumtemperatur
MDL: MFCD00072042
CAS-Nummer: 7695-91-2
EINECS: 231-710-0

Bestellen

Spezifikation Testergebnisse

Assay 96% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR