DL-α-Tocopherol 96,0-102,0%
Lieferant: Spectrum Chemical
Synonyme:
(±)-α-Tocopherol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E Alkohol, Vitamin E
YY2750-1L
SPCMYY2750-1LEA
2420
EUR
SPCMYY2750-1L
DL-α-Tocopherol 96,0-102,0%
DL-α-Tocopherol
Spectrum™ Chemical vitamin E, 1 mg = 1.1 IU, USP is used as an antioxidant. All spectrum chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₂₉H₅₀O₂ Molecular Weight: 430,71 g/mol |
MDL:
MFCD00072051 CAS-Nummer: 10191-41-0 |
Spezifikation Testergebnisse
Acidity | 1.0 ml |
Purity | 96 - 102% |
Grade | USP |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...