Zink-D-gluconat ≥97,0% USP

Lieferant: Spectrum Chemical

Synonyme: Zinc gluconate

Z1077-12KG Z1077-50KGBL Z1077-2.5KG
SPCMZ1077-12KGEA 2640 EUR
SPCMZ1077-12KG SPCMZ1077-50KGBL SPCMZ1077-2.5KG
Zink-D-gluconat ≥97,0% USP
Zink-D-gluconat

ZINC GLUCONATE, USP is a zinc salt of gluconic acid can also be manufactured by fermenting glucose. It comes as a white powder and is used in cold treating lozenges and as one of the more effective ways of delivering zinc as a dietary supplement. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₁₂H₂₂O₁₄Zn
MDL: MFCD00868110
CAS-Nummer: 4468-02-4
EINECS: 224-736-9

Bestellen

Spezifikation Testergebnisse

Assay (Anhydrous Basis) 97.0 - 102.0 %
pH of a 1 in 100 Solution 5.5 - 7.5
Water max. 11.6 %
Chloride (Cl) max. 0.05 %
Sulfate max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR