Zinkacetat Dihydrat 98,0-102,0%, Kristalle USP
Lieferant: Spectrum Chemical
|
Achtung
|
Synonyme:
Zink(II)acetat Dihydrat, Zink(II)acetat-Dihydrat, Zinkdiacetat-Dihydrat, Zinkdiacetat Dihydrat, Essigsäure Zinksalz Dihydrat, Essigsäure Zinksalz-Dihydrat
Zinc Acetate, Crystal, USP is used in anti-itch oitments and for treating zinc deficiencies. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formel:
(H₃CCOO)₂Zn·2H₂O Molecular Weight: 219.51 g/mol Schmelzpunkt: 237 Dichte: 1.718 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066961 CAS-Nummer: 5970-45-6 EINECS: 209-170-2 UN: 3077 ADR: 9 III Merck Index: 12,10256 |
Spezifikation Testergebnisse
ASSAY [Zn(CH3COO)2.2H2O] | 98.0-102.0 % |
pH OF A 5% SOLUTION @ 25°C | 6.0-8.0 |
INSOLUBLE MATTER | 0.005 % |
ARSENIC (As) | 3 ppm |
LEAD (Pb) | 0.002 % |
CHLORIDE (Cl) | 0.005 % |
SULFATES (SO4) | 0.010 % |
ALKALIES AND ALKALINE EARTHS | 0.2 % |
ELEMENTAL IMPURITIES | AS REPORTED |
IDENTIFICATION | TO PASS TEST |
RESIDUAL SOLVENTS | TO PASS TEST |
CERTIFIED HALAL |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...