Certified colour reference standards for spectrophotometers
Lieferant: Thermo Fisher Scientific
9423UV95520E 9423UV95100E 401322989841
SPET9423UV95520EEA 2360 EUR
SPET9423UV95520E SPET9423UV95100E SPET401322989841
Certified colour reference standards for spectrophotometers
Standards Farbreferenzen und Spektroskopie-Standards
These colour reference standards can be used with UV-Vis spectroscopy.
  • Each standard includes a calibration certificate
Standards: Traceable to ISO/IEC Standard 17025 accredited calibration process.