Certified colour reference standards for spectrophotometers
Lieferant: Thermo Fisher Scientific
9423UV95520E
9423UV95100E
401322989841
SPET9423UV95520EEA
2360
EUR
SPET9423UV95520E
SPET9423UV95100E
SPET401322989841
Certified colour reference standards for spectrophotometers
Standards
Farbreferenzen und Spektroskopie-Standards
These colour reference standards can be used with UV-Vis spectroscopy.
- Each standard includes a calibration certificate
Standards: Traceable to ISO/IEC Standard 17025 accredited calibration process.