Molecular model: DNA, Molymod®
Lieferant: SPIRING ENTERPRISES
MKO-ACGT-53
SPIRMKO-ACGT-53EA 0 EUR
SPIRMKO-ACGT-53
Molecular model: DNA, Molymod®
Modelle Molekularbiologie
This molecular model can show how the bases (adenin, cytosin, guanin, thymin) are combined through triple and double hydrogen bonds.