Molecular model: ten-layer DNA, Molymod®
Lieferant: SPIRING ENTERPRISES
MKS-122-10
SPIRMKS-122-10EA 242 EUR
SPIRMKS-122-10
Molecular model: ten-layer DNA, Molymod®
Modelle Molekularbiologie
Molecular model of compact DNA including ten pairs of bases.
  • Height ca. 50 cm
Bestellinformation: Self-assembly kit incl. 640 atoms and 750 connections on a stand.